N-(4-Fluorophenyl)anthranilic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-[(4-Fluorophenyl)amino]benzoic acid

008962-1G 008962-5G
101795-948EA 61.54 USD
101795-948 101795-950
N-(4-Fluorophenyl)anthranilic acid ≥97%
N-(4-Fluorophenyl)anthranilic acid

Hazard: Irritant

Formula: C₁₃H₁₀FNO₂
MW: 231.22 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD01675229
CAS Number: 54-60-4

Order Now

Specification Test Results

Melting point 198-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR