6-Fluoro-6-deoxy-D-galactopyranose ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Deoxy-6-fluoro-D-galactopyranose

008942-25MG 008942-100MG
101795-876EA 170.77 USD
101795-876 101795-878
6-Fluoro-6-deoxy-D-galactopyranose ≥98%
6-Fluoro-6-deoxy-D-galactopyranose

Hazard: Irritant/Irritant-Harmful

Formula: C₆H₁₁FO₅
MW: 182.15 g/mol
Melting Pt: 161…162.5 °C
MDL Number: MFCD00063304
CAS Number: 18961-68-7

Order Now

Specification Test Results

Melting point 161-162.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR