2-Fluoroadenine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-fluoropurine, 6-Amino-2-fluoro-7H-purine, 2-Fluoro-7H-purin-6-amine

008939-1G 008939-5G
101795-866EA 61.54 USD
101795-866 101795-868
2-Fluoroadenine ≥97%
2-Fluoroadenine

Hazard: Irritant/Irritant-Harmful

Formula: C₅H₄FN₅
MW: 153.12 g/mol
Melting Pt: >350 °C
MDL Number: MFCD01632749
CAS Number: 700-49-2

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR