2-Bromo-4,5,6-trichloroaniline ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2,3,4-trifluoroaniline, 2-Bromo-4,5,6-trifluoroaniline

008856-1G 008856-5G
101795-632EA 24.62 USD
101795-632 101795-634
2-Bromo-4,5,6-trichloroaniline ≥98%
2-Bromo-4,5,6-trichloroaniline

Hazard: Irritant

Formula: C₆H₃BrF₃N
MW: 225.9900 g/mol
Melting Pt: 58-60°C
MDL Number: MFCD00013388
CAS Number: 122375-82-0

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR