5-Bromo-2,4-difluoroaniline ≥98%

Supplier: Matrix Scientific

008831-25G 008831-100G
101795-556EA 83.08 USD
101795-556 101795-558
5-Bromo-2,4-difluoroaniline ≥98%
5-Bromo-2,4-difluoroaniline

Hazard: Irritant/Store in Freezer

Formula: C₆H₄BrF₂N
Melting Pt: 25…26 °C
MDL Number: MFCD03426368
CAS Number: 452-92-6

Order Now

Specification Test Results

Melting point 25-26°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR