6-Bromo-4-chloro-2-(trifluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

008826-250MG 008826-1G
101795-540EA 113.85 USD
101795-540 101795-542
6-Bromo-4-chloro-2-(trifluoromethyl)quinoline ≥97%
6-Bromo-4-chloro-2-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₄BrClF₃N
MW: 310.51 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD00153073
CAS Number: 1701-28-6

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR