Bis(1H,1H-perfluorobutyl)fumarate ≥97%

Supplier: Matrix Scientific

008814-1G 008814-5G
101795-512EA 43.08 USD
101795-512 101795-514
Bis(1H,1H-perfluorobutyl)fumarate ≥97%
Bis(1H,1H-perfluorobutyl)fumarate

Hazard: Irritant

Formula: C₁₂H₆F₁₄O₄
MW: 480.16 g/mol
Melting Pt: 45 °C
MDL Number: MFCD00337741
CAS Number: 24120-17-0

Order Now

Specification Test Results

Melting point 40-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR