2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane ≥98%

Supplier: Matrix Scientific

Synonyms: 4,4'-(Hexafluoroisopropylidene)bis[(4-aminophenoxy)benzene]

008801-25G 008801-100G
101795-480EA 116.92 USD
101795-480 101795-482
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane ≥98%
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane

Hazard: Irritant

Formula: C₂₇H₂₀F₆N₂O₂
MW: 518.46 g/mol
Melting Pt: 163 °C
MDL Number: MFCD00015723
CAS Number: 69563-88-8

Order Now

Specification Test Results

Melting point 163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR