3-Nitro-5-(trifluoromethyl)anisole ≥99%
Supplier: Matrix Scientific
Synonyms:
3-Methoxy-5-nitrobenzotrifluoride
008745-5G
008745-25G
101795-348EA
43.08
USD
101795-348
101795-350
3-Nitro-5-(trifluoromethyl)anisole ≥99%
3-Nitro-5-(trifluoromethyl)anisole
Hazard: Irritant
Formula:
C₈H₆F₃NO₃ MW: 221.13 g/mol Melting Pt: 35°C |
MDL Number:
MFCD00010446 CAS Number: 328-79-0 |
Specification Test Results
Melting point | 35°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...