2-Bromo-3,4,6-trifluoroaniline ≥99%

Supplier: Matrix Scientific

008708-5G 008708-25G
101795-242EA 156.92 USD
101795-242 101795-244
2-Bromo-3,4,6-trifluoroaniline ≥99%
2-Bromo-3,4,6-trifluoroaniline

Hazard: Irritant

Formula: C₆H₃BrF₃N
MW: 225.993 g/mol
Melting Pt: 36-37°C°C
MDL Number: MFCD00012370
CAS Number: 1481-21-6

Order Now

Specification Test Results

Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR