{1-Methyl-3-phenyl-1H-thieno[2,3-c]pyrazol-5-yl}methanol ≥95%

Supplier: Matrix Scientific

059321-500MG 059321-1G 059321-5G
101917-902EA 992.31 USD
101917-902 101917-904 101917-906
{1-Methyl-3-phenyl-1H-thieno[2,3-c]pyrazol-5-yl}methanol ≥95%
{1-Methyl-3-phenyl-1H-thieno[2,3-c]pyrazol-5-yl}methanol

Hazard: Irritant

Formula: C₁₃H₁₂N₂OS
Melting Pt: 135…137 °C
MDL Number: MFCD18426519
CAS Number: 1263063-15-5

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR