1-Methyl-5-nitro-1H-1,2,3-benzotriazole ≥95%

Supplier: Matrix Scientific

059320-500MG 059320-1G
101917-898EA 346.15 USD
101917-898 101917-900
1-Methyl-5-nitro-1H-1,2,3-benzotriazole ≥95%
1-Methyl-5-nitro-1H-1,2,3-benzotriazole

Hazard: Irritant

Formula: C₇H₆N₄O₂
Melting Pt: 162…164 °C
MDL Number: MFCD00972294
CAS Number: 25877-34-3

Order Now

Specification Test Results

Melting point 162-164°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR