Methyl N-(cis)-1-benzyl-4-methylpiperidin-3-yl]carbamate hydrochloride ≥95%

Supplier: Matrix Scientific

059316-500MG 059316-1G
101917-880EA 992.31 USD
101917-880 101917-882
Methyl N-(cis)-1-benzyl-4-methylpiperidin-3-yl]carbamate hydrochloride ≥95%
Methyl N-(cis)-1-benzyl-4-methylpiperidin-3-yl]carbamate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MW: 298.82 g/mol
Melting Pt: 207 °C (dec.)
MDL Number: MFCD11044502
CAS Number: 694495-65-3

Order Now

Specification Test Results

Melting point 207°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR