Methyl-2-amino-5-hydroxy-4-methoxybenzoate ≥95%

Supplier: Matrix Scientific

059315-500MG 059315-1G
101917-876EA 598.46 USD
101917-876 101917-878
Methyl-2-amino-5-hydroxy-4-methoxybenzoate ≥95%
Methyl-2-amino-5-hydroxy-4-methoxybenzoate

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 160 °C (dec.)
MDL Number: MFCD17168892
CAS Number: 50413-44-0

Order Now

Specification Test Results

Melting point 160°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR