5-Hydroxy-4-methoxy-2-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-hydroxy-4-methoxy-2-nitrobenzoic acid

059314-500MG 059314-1G 059314-5G
101917-870EA 315.38 USD
101917-870 101917-872 101917-874
5-Hydroxy-4-methoxy-2-nitrobenzoic acid ≥95%
5-Hydroxy-4-methoxy-2-nitrobenzoic acid

Hazard: Irritant

Formula: C₈H₇NO₆
MW: 213.15 g/mol
Melting Pt: 178…180 °C (dec.)
MDL Number: MFCD10666941
CAS Number: 31839-20-0

Order Now

Specification Test Results

Melting point 178-180°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR