4-Fluoro-3-phenyl-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

059312-500MG 059312-1G
101917-862EA 598.46 USD
101917-862 101917-864
4-Fluoro-3-phenyl-1H-pyrazol-5-amine ≥95%
4-Fluoro-3-phenyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₉H₈FN₃
Melting Pt: 109…111 °C
MDL Number: MFCD18426517
CAS Number: 1246931-19-0

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR