[(Diphenylmethyl)sulfanyl]methanimidamidehydrobromide ≥95%

Supplier: Matrix Scientific

059302-500MG 059302-1G
101917-820EA 295.38 USD
101917-820 101917-822
[(Diphenylmethyl)sulfanyl]methanimidamidehydrobromide ≥95%
[(Diphenylmethyl)sulfanyl]methanimidamidehydrobromide

Hazard: Irritant

Formula: C₁₄H₁₅BrN₂S
Melting Pt: 178…180 °C
MDL Number: MFCD00137024
CAS Number: 90280-15-2

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR