1,2-Dimethyl-1H-1,3-benzodiazol-5-amine ≥95%

Supplier: Matrix Scientific

059299-500MG 059299-1G
101917-812EA 860 USD
101917-812 101917-814
1,2-Dimethyl-1H-1,3-benzodiazol-5-amine ≥95%
1,2-Dimethyl-1H-1,3-benzodiazol-5-amine

Hazard: Irritant

Formula: C₉H₁₁N₃
Melting Pt: 154…156 °C
MDL Number: MFCD03872210
CAS Number: 3527-19-3

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR