3-(4-Chlorophenyl)-4-fluoro-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

059296-500MG 059296-1G
101917-804EA 598.46 USD
101917-804 101917-806
3-(4-Chlorophenyl)-4-fluoro-1H-pyrazol-5-amine ≥95%
3-(4-Chlorophenyl)-4-fluoro-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₉H₇ClFN₃
Melting Pt: 163…165 °C
MDL Number: MFCD18426511
CAS Number: 1246384-94-0

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR