3-(4-Chlorophenyl)-2-fluoro-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

059295-500MG 059295-1G
101917-800EA 401.54 USD
101917-800 101917-802
3-(4-Chlorophenyl)-2-fluoro-3-oxopropanenitrile ≥95%
3-(4-Chlorophenyl)-2-fluoro-3-oxopropanenitrile

Hazard: Irritant

Formula: C₉H₅ClFNO
Melting Pt: 58…60 °C
MDL Number: MFCD18426510
CAS Number: 1263063-13-3

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR