2-(Chloromethyl)-1-methyl-5-nitro-1H-1,3-benzodiazole ≥95%

Supplier: Matrix Scientific

059293-500MG 059293-1G
101917-792EA 307.69 USD
101917-792 101917-794
2-(Chloromethyl)-1-methyl-5-nitro-1H-1,3-benzodiazole ≥95%
2-(Chloromethyl)-1-methyl-5-nitro-1H-1,3-benzodiazole

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
Melting Pt: 193…195 °C
MDL Number: MFCD01073119
CAS Number: 20443-39-4

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR