2-Chloro-N-methoxy-N-methylpyridine-3-carboxamide ≥95%

Supplier: Matrix Scientific

059292-500MG 059292-1G 059292-5G
101917-786EA 295.38 USD
101917-786 101917-788 101917-790
2-Chloro-N-methoxy-N-methylpyridine-3-carboxamide ≥95%
2-Chloro-N-methoxy-N-methylpyridine-3-carboxamide

Hazard: Irritant

Formula: C₈H₉ClN₂O₂
Melting Pt: 85…87 °C
MDL Number: MFCD11847245
CAS Number: 488149-34-4

Order Now

Specification Test Results

Melting point 85-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR