4-Chloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

059290-500MG 059290-1G
101917-778EA 600 USD
101917-778 101917-780
4-Chloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine ≥95%
4-Chloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₆H₃ClFN₃
Melting Pt: 177…180 °C (dec.)
MDL Number: MFCD12924542
CAS Number: 582313-57-3

Order Now

Specification Test Results

Melting point 177-180°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR