2-Aminoquinolin-4-ol ≥95%

Supplier: Matrix Scientific

059288-500MG 059288-1G 059288-5G
101917-766EA 184.62 USD
101917-766 101917-768 101917-770
2-Aminoquinolin-4-ol ≥95%
2-Aminoquinolin-4-ol

Hazard: Irritant

Formula: C₉H₈N₂O
Melting Pt: 288…290 °C (dec.)
MDL Number: MFCD00052386
CAS Number: 42712-64-1

Order Now

Specification Test Results

Melting point 288-290°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR