2,2,2-Trifluoro-N-(2-hydroxy-4-nitrophenyl)acetamide ≥95%

Supplier: Matrix Scientific

058695-500MG 058695-1G 058695-5G
101915-448EA 193.85 USD
101915-448 101915-450 101915-452
2,2,2-Trifluoro-N-(2-hydroxy-4-nitrophenyl)acetamide ≥95%
2,2,2-Trifluoro-N-(2-hydroxy-4-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₈H₅F₃N₂O₄
MW: 250.14 g/mol
Melting Pt: 166…168 °C
MDL Number: MFCD00224095
CAS Number: 785-27-3

Order Now

Specification Test Results

Melting point 166-168°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR