1-Thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

058694-500MG 058694-1G
101915-444EA 295.38 USD
101915-444 101915-446
1-Thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%
1-Thia-4-azaspiro[4.5]decane-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₂S
Melting Pt: 177…179 °C
MDL Number: MFCD00553455
CAS Number: 66065-60-9

Order Now

Specification Test Results

Melting point 177-179°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR