tert-Butyl-4-(4-fluoro-5-methyl-2-nitrophenyl)piperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
058693-500MG
058693-1G
058693-5G
101915-438EA
401.54
USD
101915-438
101915-440
101915-442
tert-Butyl-4-(4-fluoro-5-methyl-2-nitrophenyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(4-fluoro-5-methyl-2-nitrophenyl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₂FN₃O₄ Melting Pt: 113…115 °C |
MDL Number:
MFCD18157680 CAS Number: 1261079-70-2 |
Specification Test Results
Melting point | 113-115°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...