2-(Pyrimidin-2-yl)pyrimidine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

058691-500MG 058691-1G
101915-430EA 523.08 USD
101915-430 101915-432
2-(Pyrimidin-2-yl)pyrimidine-5-carboxylic acid ≥95%
2-(Pyrimidin-2-yl)pyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₉H₆N₄O₂
Melting Pt: 300…301 °C
MDL Number: MFCD16869027
CAS Number: 933191-25-4

Order Now

Specification Test Results

Melting point 300-301°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR