2-[4-(2-Methylpropoxy)phenyl]acetohydrazide ≥95%

Supplier: Matrix Scientific

058686-500MG 058686-1G 058686-5G
101915-408EA 401.54 USD
101915-408 101915-410 101915-412
2-[4-(2-Methylpropoxy)phenyl]acetohydrazide ≥95%
2-[4-(2-Methylpropoxy)phenyl]acetohydrazide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
Melting Pt: 116…118 °C
MDL Number: MFCD18157667
CAS Number: 61904-59-4

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR