2-[4-(2-Methylpropoxy)phenyl]acetic acid ≥95%

Supplier: Matrix Scientific

058684-500MG 058684-1G 058684-5G
101915-396EA 401.54 USD
101915-396 101915-398 101915-400
2-[4-(2-Methylpropoxy)phenyl]acetic acid ≥95%
2-[4-(2-Methylpropoxy)phenyl]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₃
Melting Pt: 87…89 °C
MDL Number: MFCD11131689
CAS Number: 13362-94-2

Order Now

Specification Test Results

Melting point 87-89°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR