6-Methyl-1-(piperidin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one hydrochloride ≥95%
Supplier: Matrix Scientific
058683-500MG
058683-1G
101915-392EA
553.85
USD
101915-392
101915-394
6-Methyl-1-(piperidin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one hydrochloride ≥95%
6-Methyl-1-(piperidin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClN₃O Melting Pt: 204 °C (dec.) |
MDL Number:
MFCD18157652 CAS Number: 374730-72-0 |
Specification Test Results
Melting point | 204°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...