1-Methyl-6-nitro-1,2,3,4-tetrahydroquinoxaline-2,3-dione ≥95%

Supplier: Matrix Scientific

058682-500MG 058682-1G
101915-388EA 598.46 USD
101915-388 101915-390
1-Methyl-6-nitro-1,2,3,4-tetrahydroquinoxaline-2,3-dione ≥95%
1-Methyl-6-nitro-1,2,3,4-tetrahydroquinoxaline-2,3-dione

Hazard: Irritant

Formula: C₉H₇N₃O₄
Melting Pt: >300 °C
MDL Number: MFCD00956877
CAS Number: 90323-30-1

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR