1-Methyl-5-nitro-1H-1,3-benzodiazol-2-amine hydrobromide ≥95%

Supplier: Matrix Scientific

058678-500MG 058678-1G
101915-368EA 598.46 USD
101915-368 101915-370
1-Methyl-5-nitro-1H-1,3-benzodiazol-2-amine hydrobromide ≥95%
1-Methyl-5-nitro-1H-1,3-benzodiazol-2-amine hydrobromide

Hazard: Irritant

Formula: C₈H₉BrN₄O₂
Melting Pt: 290…292 °C
MDL Number: MFCD18157675
CAS Number: 1261079-64-4

Order Now

Specification Test Results

Melting point 290-292°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR