Methyl-N-(4-methylpyridin-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

058675-500MG 058675-1G 058675-5G
101915-354EA 218.46 USD
101915-354 101915-356 101915-358
Methyl-N-(4-methylpyridin-3-yl)carbamate ≥95%
Methyl-N-(4-methylpyridin-3-yl)carbamate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
Melting Pt: 117…118 °C
MDL Number: MFCD09030045
CAS Number: 694495-63-1

Order Now

Specification Test Results

Melting point 117-118°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR