Methyl-3-methoxy-4-[(1-methylpiperidin-4-yl)methoxy]benzoate ≥95%

Supplier: Matrix Scientific

058673-500MG 058673-1G
101915-346EA 401.54 USD
101915-346 101915-348
Methyl-3-methoxy-4-[(1-methylpiperidin-4-yl)methoxy]benzoate ≥95%
Methyl-3-methoxy-4-[(1-methylpiperidin-4-yl)methoxy]benzoate

Hazard: Irritant

Formula: C₁₆H₂₃NO₄
Melting Pt: 83…85 °C
MDL Number: MFCD18157679
CAS Number: 635678-09-0

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR