Methyl-3-hydroxy-4-methoxy-2,6-dinitrobenzoate ≥95%

Supplier: Matrix Scientific

058671-500MG 058671-1G 058671-5G
101915-336EA 295.38 USD
101915-336 101915-338 101915-340
Methyl-3-hydroxy-4-methoxy-2,6-dinitrobenzoate ≥95%
Methyl-3-hydroxy-4-methoxy-2,6-dinitrobenzoate

Hazard: Irritant

Formula: C₉H₈N₂O₈
Melting Pt: 163…165 °C
MDL Number: MFCD18157646
CAS Number: 1261079-62-2

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR