Methyl-2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate

058669-1G 058669-5G
101915-328EA 276.92 USD
101915-328 101915-330
Methyl-2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate ≥95%
Methyl-2-(2-(((benzyloxy)carbonyl)amino)propan-2-yl)-5-hydroxy-6-oxo-1,6-dihydropyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₉N₃O₆
MW: 361.3493 g/mol
Melting Pt: 174…176 °C
MDL Number: MFCD12026362
CAS Number: 519032-08-7

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR