[4-(Methylamino)-3-nitrophenyl]methanol ≥95%

Supplier: Matrix Scientific

058668-500MG 058668-1G
101915-324EA 401.54 USD
101915-324 101915-326
[4-(Methylamino)-3-nitrophenyl]methanol ≥95%
[4-(Methylamino)-3-nitrophenyl]methanol

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
Melting Pt: 125…127 °C
MDL Number: MFCD16689902
CAS Number: 62347-97-1

Order Now

Specification Test Results

Melting point 125-127°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR