4-(Methylamino)-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

058667-500MG 058667-1G
101915-320EA 295.38 USD
101915-320 101915-322
4-(Methylamino)-3-nitrobenzaldehyde ≥95%
4-(Methylamino)-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₈H₈N₂O₃
Melting Pt: 173…175 °C
MDL Number: MFCD03964545
CAS Number: 42564-41-0

Order Now

Specification Test Results

Melting point 173-175°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR