5-Iodoquinazoline-2,4-diamine ≥95%

Supplier: Matrix Scientific

058665-500MG 058665-1G 058665-5G
101915-310EA 261.54 USD
101915-310 101915-312 101915-314
5-Iodoquinazoline-2,4-diamine ≥95%
5-Iodoquinazoline-2,4-diamine

Hazard: Irritant

Formula: C₈H₇IN₄
Melting Pt: 201…203 °C
MDL Number: MFCD01411818
CAS Number: 119584-76-8

Order Now

Specification Test Results

Melting point 201-203°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR