2-(4-Fluoro-5-methyl-2-nitrophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

058659-500MG 058659-1G
101915-284EA 401.54 USD
101915-284 101915-286
2-(4-Fluoro-5-methyl-2-nitrophenyl)acetic acid ≥95%
2-(4-Fluoro-5-methyl-2-nitrophenyl)acetic acid

Hazard: Irritant

Formula: C₉H₈FNO₄
Melting Pt: 135…137 °C
MDL Number: MFCD18157661
CAS Number: 1261079-63-3

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR