1-Fluoro-4-methoxy-2-methyl-5-nitrobenzene ≥95%
Supplier: Matrix Scientific
058658-500MG
058658-1G
101915-280EA
401.54
USD
101915-280
101915-282
1-Fluoro-4-methoxy-2-methyl-5-nitrobenzene ≥95%
1-Fluoro-4-methoxy-2-methyl-5-nitrobenzene
Hazard: Irritant
Formula:
C₈H₈FNO₃ Melting Pt: 79…81 °C |
MDL Number:
MFCD18157656 CAS Number: 182880-71-3 |
Specification Test Results
Melting point | 79-81°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...