Ethyl-6-oxo-2-(pyrimidin-2-yl)-1,6-dihydropyrimidine-5-carboxylate ≥95%

Supplier: Matrix Scientific

058655-500MG 058655-1G
101915-268EA 401.54 USD
101915-268 101915-270
Ethyl-6-oxo-2-(pyrimidin-2-yl)-1,6-dihydropyrimidine-5-carboxylate ≥95%
Ethyl-6-oxo-2-(pyrimidin-2-yl)-1,6-dihydropyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₄O₃
Melting Pt: 166…168 °C
MDL Number: MFCD15511375
CAS Number: 432521-54-5

Order Now

Specification Test Results

Melting point 166-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR