Ethyl-3-iodo-1H-indole-2-carboxylate ≥95%
Supplier: Matrix Scientific
058654-500MG
058654-1G
058654-5G
101915-262EA
193.85
USD
101915-262
101915-264
101915-266
Ethyl-3-iodo-1H-indole-2-carboxylate ≥95%
Ethyl-3-iodo-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀INO₂ Melting Pt: 136…137 °C |
MDL Number:
MFCD18157645 CAS Number: 117637-79-3 |
Specification Test Results
Melting point | 136-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...