Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

058653-500MG 058653-1G 058653-5G
101915-256EA 295.38 USD
101915-256 101915-258 101915-260
Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate hydrochloride ≥95%
Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₀ClIN₂O₂
Melting Pt: 191…192 °C
MDL Number: MFCD18157674
CAS Number: 1261079-76-8

Order Now

Specification Test Results

Melting point 191-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR