Ethyl imidazo[1,2-a]pyridine-7-carboxylate ≥95%

Supplier: Matrix Scientific

058652-500MG 058652-1G 058652-5G
101915-250EA 295.38 USD
101915-250 101915-252 101915-254
Ethyl imidazo[1,2-a]pyridine-7-carboxylate ≥95%
Ethyl imidazo[1,2-a]pyridine-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.1986 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD08436046
CAS Number: 372147-49-4

Order Now

Specification Test Results

Melting point 87-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR