Ethyl-4-chloro-2-(pyrimidin-2-yl)pyrimidine-5-carboxylate ≥95%

Supplier: Matrix Scientific

058649-500MG 058649-1G
101915-236EA 804.62 USD
101915-236 101915-238
Ethyl-4-chloro-2-(pyrimidin-2-yl)pyrimidine-5-carboxylate ≥95%
Ethyl-4-chloro-2-(pyrimidin-2-yl)pyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₄O₂
Melting Pt: 158…160 °C
MDL Number: MFCD18157672
CAS Number: 432521-55-6

Order Now

Specification Test Results

Melting point 158-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR