Ethyl 2-chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate ≥95%

Supplier: Matrix Scientific

058646-500MG 058646-1G 058646-5G
101915-224EA 193.85 USD
101915-224 101915-226 101915-228
Ethyl 2-chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate ≥95%
Ethyl 2-chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
Melting Pt: 94 °C
MDL Number: MFCD00446053
CAS Number: 27143-07-3

Order Now

Specification Test Results

Melting point 104-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR