2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetaldehydeyl-4,6-dichloropyrimidine

058641-1G 058641-5G
101915-208EA 83.08 USD
101915-208 102833-268
2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde ≥95%
2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde

Hazard: Irritant/Store in Freezer

Formula: C₆H₄Cl₂N₂O
MW: 191.0200 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD09991563
CAS Number: 16019-33-3

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR