5,8-Dichloro-7-nitro-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

058640-500MG 058640-1G
101915-202EA 992.31 USD
101915-202 101915-204
5,8-Dichloro-7-nitro-1,2,3,4-tetrahydroisoquinoline ≥95%
5,8-Dichloro-7-nitro-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₉H₈Cl₂N₂O₂
Melting Pt: 128…130 °C
MDL Number: MFCD18157653
CAS Number: 1261079-69-9

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR